Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018420.2 | NP_060890.2 | 8 | 1267 | ||
GenBank mRNA | AK125072.1 | - | 6 | 835 | ||
AY145501.1 | - | 8 | 1137 | |||
BC026358.1 | - | 8 | 1266 | 4814675 | ||
KU178698.1 | - | 8 | 1101 | |||
KU178699.1 | - | 9 | 1432 | |||
KU178700.1 | - | 8 | 1101 | |||
KU178701.1 | - | 10 | 1219 | |||
KU178702.1 | - | 8 | 1024 | |||
KU178703.1 | - | 7 | 888 |