Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000707.3 | NP_000698.1 | 1 | 955 | ||
GenBank mRNA | AF030512.1 | - | 1 | 920 | ||
AF101726.1 | - | 1 | 920 | |||
D31833.1 | - | 1 | 1004 | |||
EU432111.1 | - | 1 | 797 | |||
L37112.1 | - | 1 | 891 |
Set Membership: |
G-protein coupled receptor |