Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291780.1 | NP_001278709.1 | 3 | 599 | ||
NM_017495.5 | NP_059965.2 | 2 | 503 | |||
NM_183425.2 | NP_906270.1 | 2 | 503 | |||
XM_005260446.3 | XP_005260503.1 | 3 | 537 | |||
XM_011528885.2 | XP_011527187.1 | 3 | 531 | |||
GenBank mRNA | AF432218.1 | - | 2 | 499 | ||
BC018711.1 | - | 2 | 255 | 4852523 | ||
BE840212.1 | - | 3 | 228 | |||
BF371746.1 | - | 3 | 234 | |||
BM549757.1 | - | 498 | ||||
DN998561.1 | - | 2 | 488 | |||
X75314.1 | - | 2 | 290 |
Gene Symbol: LOC100291105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096426.1 | - | 1 | 324 |