Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF21A SIRNA Assays ›
Gene Symbol:
KIF21A
Gene Name:
kinesin family member 21A
Gene Aliases:
CFEOM1, FEOM1, FEOM3A
Chromosome Location:
Chr.12: 39293228 - 39443414 on Build GRCh38
UniGene ID:
Hs.374201
Species:
Human
siRNA ID
HSS183140
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIF21A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173463.1
NP_001166934.1
21
3518
NM_001173464.1
NP_001166935.1
22
3557
NM_001173465.1
NP_001166936.1
20
3449
NM_017641.3
NP_060111.2
21
3518
XM_005269007.2
XP_005269064.1
22
3581
XM_005269008.2
XP_005269065.1
22
3581
XM_005269009.2
XP_005269066.1
22
3581
XM_005269010.2
XP_005269067.1
21
3542
XM_005269011.2
XP_005269068.1
22
3581
XM_005269012.2
XP_005269069.1
22
3581
XM_005269013.2
XP_005269070.1
22
3581
XM_005269014.2
XP_005269071.1
22
3581
XM_006719493.2
XP_006719556.1
21
3542
XM_006719494.2
XP_006719557.1
22
3581
XM_011538556.2
XP_011536858.1
21
3512
XM_017019607.1
XP_016875096.1
21
3542
XM_017019608.1
XP_016875097.1
21
3542
XM_017019609.1
XP_016875098.1
21
3542
XM_017019610.1
XP_016875099.1
22
3581
XM_017019611.1
XP_016875100.1
21
3542
GenBank mRNA
AB051495.2
-
21
3087
AB290166.1
-
21
3518
AF450487.1
-
21
3247
AM177179.1
-
20
3029
AY368076.1
-
22
3153
BC136414.1
-
21
3187
9020402
BX537855.1
-
8
1180
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0