Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017528.2 | NP_001017528.1 | 8 | 864 | ||
NM_001017529.2 | NP_001017529.1 | 7 | 747 | |||
NM_001017530.1 | NP_001017530.1 | 7 | 921 | |||
NM_001198721.1 | NP_001185650.1 | 9 | 1577 | |||
NM_015366.3 | NP_056181.2 | 8 | 1038 | |||
NM_181333.3 | NP_851850.1 | 7 | 899 | |||
GenBank mRNA | AF177331.1 | - | 7 | 843 | ||
AK000192.1 | - | 3 | 957 | |||
AK055848.1 | - | 9 | 1300 | |||
AK056787.1 | - | 1 | 985 | |||
AK091884.1 | - | 8 | 789 | |||
AK291564.1 | - | 7 | 889 | |||
BC016921.1 | - | 3 | 1146 | 4413787 |
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL355192.1 | - | 7 | 622 |