Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171083.2 | NP_001164554.1 | 5 | 521 | ||
NM_017751.4 | NP_060221.2 | 7 | 740 | |||
NM_017951.4 | NP_060421.2 | 7 | 740 | |||
NR_033230.2 | 7 | 1733 | ||||
NR_033231.2 | 6 | 597 | ||||
NR_033232.2 | 6 | 664 | ||||
XM_011511443.2 | XP_011509745.1 | 7 | 902 | |||
XM_011511444.2 | XP_011509746.1 | 7 | 901 | |||
XM_011511445.2 | XP_011509747.1 | 7 | 596 | |||
XM_017004448.1 | XP_016859937.1 | 7 | 642 | |||
XM_017004449.1 | XP_016859938.1 | 7 | 642 | |||
GenBank mRNA | AB037839.1 | - | 7 | 1733 | ||
AK000304.1 | - | 7 | 577 | |||
AK000763.1 | - | 2 | 70 | |||
AK056501.1 | - | 7 | 532 | |||
AK299185.1 | - | 5 | 371 | |||
AK303519.1 | - | 6 | 501 | |||
BC064947.1 | - | 6 | 419 | 6023304 | ||
EU367941.1 | - | 7 | 584 |