Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171083.2 | NP_001164554.1 | 7 | 784 | ||
NM_017751.4 | NP_060221.2 | 9 | 1003 | |||
NM_017951.4 | NP_060421.2 | 9 | 1003 | |||
NR_033230.2 | 9 | 1996 | ||||
NR_033231.2 | 8 | 860 | ||||
NR_033232.2 | 8 | 927 | ||||
XM_011511443.2 | XP_011509745.1 | 9 | 1165 | |||
XM_011511444.2 | XP_011509746.1 | 9 | 1164 | |||
XM_011511445.2 | XP_011509747.1 | 9 | 859 | |||
XM_017004448.1 | XP_016859937.1 | 9 | 905 | |||
XM_017004449.1 | XP_016859938.1 | 9 | 905 | |||
GenBank mRNA | AB037839.1 | - | 9 | 1996 | ||
AK000304.1 | - | 9 | 840 | |||
AK299185.1 | - | 7 | 634 | |||
AK303427.1 | - | 3 | 221 | |||
BC040941.1 | - | 2 | 208 |
Gene Symbol: LOC150776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026922.1 | 9 | 1102 | |||
GenBank mRNA | AK293811.1 | - | 9 | 839 | ||
AK303293.1 | - | 7 | 672 | |||
AK304811.1 | - | 8 | 900 | |||
BC093080.1 | - | 9 | 825 |