Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017772.3 | NP_060242.2 | 8 | 1136 | ||
NR_130108.1 | 8 | 1197 | ||||
XM_011514738.2 | XP_011513040.1 | 8 | 1088 | |||
XM_011514739.1 | XP_011513041.1 | 8 | 1088 | |||
GenBank mRNA | AB449909.1 | - | 8 | 922 | ||
AK000344.1 | - | 1 | 43 | |||
AK096340.1 | - | 8 | 1066 | |||
AK292893.1 | - | 8 | 1031 | |||
AY313781.1 | - | 8 | 1068 | |||
BC047658.1 | - | 8 | 1129 | |||
BC063523.1 | - | 3 | 146 | |||
BC109026.1 | - | 8 | 1066 | 40027698 | ||
BC109027.1 | - | 8 | 1066 | 40027700 | ||
BX648317.1 | - | 8 | 1067 | |||
CN423653.1 | - | 5 | 492 | |||
CR749710.1 | - | 2 | 222 |