Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC48A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017842.2 | NP_060312.2 | 3 | 768 | ||
XM_005269016.4 | XP_005269073.1 | 4 | 783 | |||
XM_017019612.1 | XP_016875101.1 | 5 | 1004 | |||
XM_017019613.1 | XP_016875102.1 | 5 | 932 | |||
XM_017019614.1 | XP_016875103.1 | 5 | 933 | |||
XM_017019615.1 | XP_016875104.1 | 5 | 1056 | |||
XM_017019616.1 | XP_016875105.1 | 4 | 785 | |||
GenBank mRNA | AK000496.1 | - | 3 | 744 | ||
BC011823.2 | - | 3 | 594 | |||
BC026344.1 | - | 3 | 714 | 4798578 | ||
BC065033.1 | - | 3 | 716 | 6147682 |