Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017892.3 | NP_060362.3 | 17 | 1850 | ||
XM_005246676.4 | XP_005246733.3 | 17 | 2370 | |||
XM_005246677.4 | XP_005246734.2 | 17 | 1846 | |||
XM_011511446.2 | XP_011509748.1 | 16 | 2316 | |||
XM_011511447.2 | XP_011509749.1 | 17 | 2370 | |||
XM_011511448.2 | XP_011509750.1 | 16 | 1792 | |||
XM_011511449.2 | XP_011509751.1 | 19 | 2200 | |||
XM_017004453.1 | XP_016859942.1 | 17 | 2370 | |||
XM_017004454.1 | XP_016859943.1 | 16 | 2316 | |||
XM_017004455.1 | XP_016859944.1 | 16 | 1792 | |||
XM_017004456.1 | XP_016859945.1 | 18 | 2036 | |||
XM_017004457.1 | XP_016859946.1 | 19 | 2199 | |||
XM_017004458.1 | XP_016859947.1 | 17 | 1946 | |||
XM_017004459.1 | XP_016859948.1 | 17 | 2055 | |||
XM_017004460.1 | XP_016859949.1 | 17 | 1983 | |||
XM_017004461.1 | XP_016859950.1 | 17 | 2055 | |||
GenBank mRNA | AF155096.1 | - | 3 | 287 | ||
AK021621.1 | - | 2 | 938 | |||
AK298547.1 | - | 14 | 1687 |