Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017895.7 | NP_060365.7 | 14 | 1786 | ||
XM_011528888.2 | XP_011527190.1 | 15 | 1850 | |||
GenBank mRNA | AF193054.1 | - | 11 | 1343 | ||
AF336851.1 | - | 8 | 1020 | |||
AK000603.1 | - | 5 | 561 | |||
AK022979.1 | - | 14 | 1656 | |||
AK025891.1 | - | 10 | 1151 | |||
AK127128.1 | - | 1 | 2618 | |||
AK223116.1 | - | 5 | 561 | |||
AK300135.1 | - | 14 | 1665 | |||
AL512707.1 | - | 1 | 14 | |||
AL832131.1 | - | 12 | 3990 | |||
AY044431.1 | - | 14 | 1725 | |||
BC009304.1 | - | 14 | 1644 | |||
BC011927.2 | - | 14 | 1640 | |||
BC016060.1 | - | 14 | 1640 | |||
BC126287.1 | - | 14 | 1775 | 8992003 | ||
BC130275.1 | - | 14 | 1775 | 40146306 | ||
BC144125.1 | - | 14 | 1775 | 9052645 | ||
EU718675.1 | - | 14 | 1693 |