Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017895.7 | NP_060365.7 | 8 | 1002 | ||
XM_011528888.2 | XP_011527190.1 | 8 | 973 | |||
GenBank mRNA | AF193054.1 | - | 5 | 560 | ||
AF336851.1 | - | 2 | 236 | |||
AK022979.1 | - | 8 | 872 | |||
AK025891.1 | - | 4 | 367 | |||
AK300135.1 | - | 8 | 881 | |||
AL832131.1 | - | 8 | 918 | |||
AY044431.1 | - | 8 | 941 | |||
BC009304.1 | - | 8 | 860 | |||
BC011927.2 | - | 8 | 856 | |||
BC016060.1 | - | 8 | 856 | |||
BC126287.1 | - | 8 | 991 | 8992003 | ||
BC130275.1 | - | 8 | 991 | 40146306 | ||
BC144125.1 | - | 8 | 991 | 9052645 | ||
EU718675.1 | - | 8 | 909 |