Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304453.1 | NP_001291382.1 | 3 | 650 | ||
NM_017908.3 | NP_060378.1 | 4 | 991 | |||
XM_005259052.4 | XP_005259109.1 | 4 | 992 | |||
XM_005259053.4 | XP_005259110.1 | 4 | 992 | |||
XM_006723266.3 | XP_006723329.1 | 4 | 992 | |||
XM_017026949.1 | XP_016882438.1 | 4 | 992 | |||
XM_017026950.1 | XP_016882439.1 | 4 | 992 | |||
GenBank mRNA | AK000633.1 | - | 4 | 730 | ||
AK026153.1 | - | 4 | 705 | |||
AL833955.1 | - | 3 | 661 | |||
AY279351.1 | - | 4 | 730 | |||
AY280800.1 | - | 4 | 770 | |||
BC017206.2 | - | 4 | 712 | 3636172 | ||
BQ441187.1 | - | 3 | 377 |