Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304349.1 | NP_001291278.1 | 3, 4 | 877 | ||
NM_002730.3 | NP_002721.1 | 3, 4 | 443 | |||
NM_207518.2 | NP_997401.1 | 3, 4 | 252 | |||
XM_017026948.1 | XP_016882437.1 | 3, 4 | 299 | |||
GenBank mRNA | AF208004.1 | - | 3, 4 | 237 | ||
AF239744.1 | - | 3, 4 | 382 | |||
AK131561.1 | - | 3, 4 | 877 | |||
AK290147.1 | - | 4, 5 | 325 | |||
AK292284.1 | - | 3, 4 | 252 | |||
AK301264.1 | - | 3, 4 | 612 | |||
BC039846.1 | - | 3, 4 | 439 | 6066764 | ||
BC108259.1 | - | 3, 4 | 440 | 6144307 | ||
BU543573.1 | - | 443 | ||||
M80335.1 | - | 2, 3 | 192 | |||
X07767.1 | - | 3, 4 | 323 |