Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136037.2 | NP_001129509.2 | 5 | 483 | ||
NM_001161403.1 | NP_001154875.1 | 4 | 417 | |||
NM_001161404.1 | NP_001154876.1 | 4 | 707 | |||
NM_017980.4 | NP_060450.2 | 4 | 528 | |||
XM_005263710.2 | XP_005263767.1 | 3 | 200 | |||
XM_006712628.3 | XP_006712691.1 | 4 | 387 | |||
XM_011511453.1 | XP_011509755.1 | 4 | 387 | |||
GenBank mRNA | AF484961.1 | - | 3 | 251 | ||
AF520987.1 | - | 4 | 872 | |||
AF527764.1 | - | 4 | 482 | |||
AF527765.1 | - | 4 | 344 | |||
AF527766.1 | - | 4 | 384 | |||
AF527767.1 | - | 3 | 640 | |||
AK000906.1 | - | 4 | 304 | |||
AK022470.1 | - | 4 | 296 | |||
AK055363.1 | - | 4 | 872 | |||
AK091652.1 | - | 3 | 929 | |||
AK094954.1 | - | 4 | 387 | |||
AK123014.1 | - | 2 | 1269 | |||
AK297645.1 | - | 6 | 675 | |||
BC065816.1 | - | 4 | 335 | 6181259 | ||
BP221000.1 | - | 4 | 528 | |||
DC356496.1 | - | 5 | 395 | |||
DR006887.1 | - | 4 | 423 |