Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018085.4 | NP_060555.2 | 12 | 1290 | ||
GenBank mRNA | AF410465.1 | - | 12 | 1240 | ||
AK027532.1 | - | 3 | 154 | |||
AK074740.1 | - | 3 | 171 | |||
BC003604.1 | - | 1 | 26 |