Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 4 | 1544 | ||
NM_001318717.1 | NP_001305646.1 | 4 | 1465 | |||
NM_001318718.1 | NP_001305647.1 | 4 | 1445 | |||
NM_001318719.1 | NP_001305648.1 | 4 | 1423 | |||
NM_001318720.1 | NP_001305649.1 | 4 | 1417 | |||
NM_001318721.1 | NP_001305650.1 | 4 | 1392 | |||
NM_001318722.1 | NP_001305651.1 | 4 | 1413 | |||
NM_001318723.1 | NP_001305652.1 | 4 | 1497 | |||
NM_001318724.1 | NP_001305653.1 | 4 | 1497 | |||
NM_001318725.1 | NP_001305654.1 | 4 | 1400 | |||
NM_016506.6 | NP_057590.3 | 4 | 1501 | |||
NM_018095.5 | NP_060565.4 | 4 | 1396 | |||
XM_017018009.1 | XP_016873498.1 | 4 | 1380 | |||
GenBank mRNA | AK001312.1 | - | 4 | 1350 | ||
AK001749.1 | - | 4 | 1437 | |||
AK091607.1 | - | 3 | 2074 | |||
AK222550.1 | - | 4 | 1347 | |||
AK225062.1 | - | 4 | 1347 | |||
AL713735.1 | - | 3 | 1326 | |||
BC002736.2 | - | 4 | 1438 | 3628290 | ||
CR457270.1 | - | 3 | 1284 |
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143984.1 | NP_001137456.1 | 2 | 2286 | ||
NM_175732.2 | NP_783859.1 | 4 | 2369 | |||
GenBank mRNA | BC014048.1 | - | 3 | 2013 |