Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002860.3 | NP_001002860.2 | 3 | 949 | ||
NM_018167.4 | NP_060637.1 | 3 | 949 | |||
XM_011536939.2 | XP_011535241.1 | 3 | 714 | |||
GenBank mRNA | AB040958.2 | - | 3 | 878 | ||
AK001510.1 | - | 3 | 875 | |||
AK021953.1 | - | 1 | 168 | |||
BC047071.1 | - | 3 | 946 | |||
BC141850.1 | - | 3 | 890 |