Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320047.1 | NP_001306976.1 | 23, 24 | 2651 | ||
NM_001320048.1 | NP_001306977.1 | 21, 22 | 2406 | |||
NM_018186.3 | NP_060656.2 | 22, 23 | 2954 | |||
XM_005245317.4 | XP_005245374.1 | 22, 23 | 2526 | |||
XM_011509735.2 | XP_011508037.1 | 19, 20 | 2136 | |||
XM_017001722.1 | XP_016857211.1 | 22, 23 | 2526 | |||
XM_017001724.1 | XP_016857213.1 | 21, 22 | 2398 | |||
XM_017001725.1 | XP_016857214.1 | 20, 21 | 2259 | |||
GenBank mRNA | AK001568.1 | - | 21, 22 | 2326 | ||
AK023532.1 | - | 12, 13 | 1389 | |||
AK096493.1 | - | 23, 24 | 2561 | |||
AK127098.1 | - | 27, 28 | 3443 | |||
AK294569.1 | - | 18, 19 | 2097 | |||
AK310638.1 | - | 18, 19 | 2067 | |||
AL354614.1 | - | 22, 23 | 2954 | |||
BC091516.1 | - | 23, 24 | 2565 | 30389801 | ||
BC107168.1 | - | 21, 22 | 2303 | 40035373 | ||
BC107169.1 | - | 22, 23 | 2442 | 40035375 |