Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145025.1 | NP_001138497.1 | 5 | 643 | ||
NM_018217.2 | NP_060687.2 | 6 | 754 | |||
NR_026728.1 | 5 | 970 | ||||
GenBank mRNA | AK001645.1 | - | 6 | 736 | ||
AK023931.1 | - | 6 | 724 | |||
AK096016.1 | - | 4 | 664 | |||
AK293583.1 | - | 4 | 574 | |||
AK298794.1 | - | 5 | 712 | |||
AK300212.1 | - | 5 | 598 | |||
AY007088.1 | - | 3 | 322 | |||
AY358580.1 | - | 5 | 604 | |||
BC001371.2 | - | 6 | 721 | 3051376 | ||
BC016184.1 | - | 6 | 695 | 4860424 | ||
CR457266.1 | - | 6 | 654 |