Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COA1 SIRNA Assays ›
Gene Symbol:
COA1
Gene Name:
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases:
C7orf44, MITRAC15
Chromosome Location:
Chr.7: 43608452 - 43729541 on Build GRCh38
UniGene ID:
Hs.268887
Species:
Human
siRNA ID
HSS183234
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: COA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321197.1
NP_001308126.1
6, 7
713
NM_001321198.1
NP_001308127.1
6, 7
594
NM_001321199.1
NP_001308128.1
6, 7
558
NM_001321200.1
NP_001308129.1
6, 7
614
NM_001321201.1
NP_001308130.1
5, 6
486
NM_018224.3
NP_060694.2
5, 6
493
NR_047701.1
6, 7
558
NR_135580.1
6, 7
594
NR_135581.1
6, 7
594
NR_135582.1
5, 6
493
NR_135583.1
5, 6
493
XM_005249807.4
XP_005249864.1
5, 6
557
XM_006715752.2
XP_006715815.1
6, 7
525
XM_017012414.1
XP_016867903.1
5, 6
484
XM_017012415.1
XP_016867904.1
6, 7
586
XM_017012416.1
XP_016867905.1
5, 6
484
XM_017012417.1
XP_016867906.1
5, 6
484
XM_017012418.1
XP_016867907.1
5, 6
484
XM_017012419.1
XP_016867908.1
5, 6
484
XM_017012421.1
XP_016867910.1
6, 7
708
XM_017012422.1
XP_016867911.1
6, 7
597
XM_017012423.1
XP_016867912.1
6, 7
517
XM_017012424.1
XP_016867913.1
5, 6
500
XM_017012425.1
XP_016867914.1
6, 7
601
XM_017012426.1
XP_016867915.1
6, 7
720
XM_017012427.1
XP_016867916.1
7, 8
675
XM_017012428.1
XP_016867917.1
5, 6
500
XM_017012429.1
XP_016867918.1
4, 5
394
XM_017012430.1
XP_016867919.1
6, 7
586
XM_017012431.1
XP_016867920.1
5, 6
500
XM_017012432.1
XP_016867921.1
5, 6
500
GenBank mRNA
AK001665.1
-
6, 7
527
AK023007.1
-
6, 7
510
AK023053.1
-
6, 7
526
AK097203.1
-
4, 5
2024
AK129690.1
-
2, 3
1217
AK293043.1
-
6, 7
536
BC056884.1
-
6, 7
485
6159045
BG028456.1
-
515
BG334368.1
-
6, 7
503
BG548154.1
-
6, 7
550
BM918741.1
-
5, 6
466
BP377872.1
-
5, 6
492
BQ231567.1
-
5, 6
431
DA742258.1
-
5, 6
475
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
translation
mitochondrial respiratory chain complex I assembly
mitochondrial respiratory chain complex IV assembly
RNA binding
structural constituent of ribosome
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline