Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018235.2 | NP_060705.2 | 5, 6 | 723 | ||
XM_005266728.2 | XP_005266785.1 | 5, 6 | 659 | |||
XM_006722503.2 | XP_006722566.1 | 5, 6 | 520 | |||
XM_011526071.2 | XP_011524373.1 | 5, 6 | 588 | |||
XM_011526072.2 | XP_011524374.1 | 5, 6 | 1543 | |||
XM_011526073.2 | XP_011524375.1 | 4, 5 | 556 | |||
GenBank mRNA | AJ347717.1 | - | 4, 5 | 487 | ||
AK001692.1 | - | 5, 6 | 626 | |||
AK024471.1 | - | 1, 2 | 92 | |||
AK097155.1 | - | 5, 6 | 738 | |||
AK298309.1 | - | 5, 6 | 342 | |||
AK300904.1 | - | 5, 6 | 590 | |||
BC001375.2 | - | 5, 6 | 596 | 3051369 | ||
BC003176.1 | - | 4, 5 | 520 | 2957870 | ||
EU794600.1 | - | 4, 5 | 520 |