Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282959.1 | NP_001269888.1 | 20, 21 | 3033 | ||
NM_001282960.1 | NP_001269889.1 | 20, 21 | 3027 | |||
NM_018237.3 | NP_060707.2 | 21, 22 | 3072 | |||
NR_104262.1 | 20, 21 | 2867 | ||||
GenBank mRNA | AF465616.1 | - | 21, 22 | 3002 | ||
AK299307.1 | - | 20, 21 | 2932 | |||
AY249140.1 | - | 21, 22 | 2980 | |||
BC130626.1 | - | 21, 22 | 2961 | 40146657 | ||
BC132725.1 | - | 21, 22 | 2961 | 40146747 | ||
BC144118.1 | - | 20, 21 | 2756 | |||
BM905703.1 | - | 449 | ||||
CB136079.1 | - | 1, 2 | 46 |