Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306147.1 | NP_001293076.1 | 7 | 1047 | ||
NM_018243.3 | NP_060713.1 | 5 | 827 | |||
XM_006714259.3 | XP_006714322.1 | 5 | 693 | |||
XM_017008400.1 | XP_016863889.1 | 6 | 836 | |||
XM_017008401.1 | XP_016863890.1 | 6 | 848 | |||
XM_017008402.1 | XP_016863891.1 | 6 | 848 | |||
XM_017008403.1 | XP_016863892.1 | 5 | 788 | |||
XM_017008404.1 | XP_016863893.1 | 5 | 788 | |||
GenBank mRNA | AK001711.1 | - | 5 | 730 | ||
AK027279.1 | - | 3 | 404 | |||
AK027633.1 | - | 2 | 242 | |||
AK094473.1 | - | 50 | ||||
AK302700.1 | - | 7 | 924 | |||
AL110300.1 | - | 5 | 709 | |||
BC008083.2 | - | 5 | 813 | 2966709 | ||
BC063615.1 | - | 5 | 678 | 3848367 | ||
GU727629.1 | - | 5 | 639 |