Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008744.1 | NP_001008744.1 | 15 | 1735 | ||
NM_018319.3 | NP_060789.2 | 16 | 1958 | |||
XM_005267847.2 | XP_005267904.1 | 16 | 2632 | |||
XM_005267848.2 | XP_005267905.1 | 15 | 1976 | |||
XM_006720197.3 | XP_006720260.1 | 16 | 2199 | |||
XM_006720198.3 | XP_006720261.1 | 15 | 1854 | |||
XM_006720199.1 | XP_006720262.1 | 15 | 1340 | |||
XM_011536942.2 | XP_011535244.1 | 15 | 2375 | |||
XM_011536943.2 | XP_011535245.1 | 15 | 1768 | |||
XM_017021439.1 | XP_016876928.1 | 16 | 1866 | |||
XM_017021442.1 | XP_016876931.1 | 13 | 1584 | |||
XM_017021443.1 | XP_016876932.1 | 13 | 1083 | |||
XM_017021444.1 | XP_016876933.1 | 13 | 1084 | |||
XM_017021446.1 | XP_016876935.1 | 14 | 1390 | |||
GenBank mRNA | AF182003.1 | - | 6 | 404 | ||
AK001952.1 | - | 15 | 1699 | |||
AK023514.1 | - | 5 | 321 | |||
AK093235.1 | - | 17 | 2766 | |||
AK315549.1 | - | 15 | 1683 | |||
AL832288.1 | - | 7 | 677 | |||
BC006083.1 | - | 18 | 2122 | |||
BC015474.1 | - | 16 | 1909 | 3900062 | ||
BC107877.1 | - | 17 | 2008 | 6597752 | ||
BX161451.1 | - | 15 | 1870 |