Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL3 SIRNA Assays ›
Gene Symbol:
MBNL3
Gene Name:
muscleblind like splicing regulator 3
Gene Aliases:
CHCR, MBLX, MBLX39, MBXL
Chromosome Location:
Chr.X: 132369314 - 132490030 on Build GRCh38
UniGene ID:
Hs.105134
Species:
Human
siRNA ID
HSS183278
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170701.1
NP_001164172.1
5
817
NM_001170702.1
NP_001164173.1
5
817
NM_001170703.1
NP_001164174.1
5
750
NM_001170704.1
NP_001164175.1
6
1017
NM_018388.3
NP_060858.2
5
980
NM_133486.2
NP_597846.1
5
875
XM_005262431.3
XP_005262488.1
6
1792
XM_005262434.3
XP_005262491.1
6
1787
XM_005262436.3
XP_005262493.1
5
1680
XM_005262437.3
XP_005262494.1
5
818
XM_005262438.3
XP_005262495.1
5
818
XM_005262439.3
XP_005262496.1
5
750
XM_005262440.3
XP_005262497.1
7
1837
XM_005262441.3
XP_005262498.1
5
941
XM_005262442.3
XP_005262499.1
5
912
XM_011531364.2
XP_011529666.1
4
1828
XM_017029632.1
XP_016885121.1
6
9222
XM_017029633.1
XP_016885122.1
7
1957
XM_017029634.1
XP_016885123.1
6
9222
XM_017029635.1
XP_016885124.1
6
9222
XM_017029636.1
XP_016885125.1
6
1721
XM_017029637.1
XP_016885126.1
6
9222
XM_017029638.1
XP_016885127.1
7
1855
XM_017029639.1
XP_016885128.1
5
930
GenBank mRNA
AB077698.1
-
5
980
AB077699.1
-
5
875
AF467070.1
-
5
875
AF491305.1
-
5
901
AJ427918.1
-
5
980
AJ427919.1
-
5
875
AK002178.1
-
5
786
AK300248.1
-
5
749
AL133625.1
-
4
725
AY072692.1
-
5
980
AY372211.1
-
5
751
BC042090.1
-
6
1016
5761647
BC074775.2
-
5
943
30915333
BC074776.2
-
5
943
30915555
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
multicellular organism development
RNA splicing
regulation of RNA splicing
negative regulation of myoblast differentiation
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0