Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290185.1 | NP_001277114.1 | 6, 7 | 1369 | ||
NM_021729.5 | NP_068375.3 | 6, 7 | 1154 | |||
XM_011542898.1 | XP_011541200.1 | 3, 4 | 387 | |||
GenBank mRNA | AB027508.2 | - | 6, 7 | 1124 | ||
AF258558.1 | - | 4, 5 | 655 | |||
AF308800.1 | - | 6, 7 | 1088 | |||
AK023279.1 | - | 6, 7 | 1125 | |||
AK056701.1 | - | 5, 6 | 981 | |||
AK223158.1 | - | 6, 7 | 1114 | |||
AK302973.1 | - | 6, 7 | 1301 | |||
AL834218.1 | - | 6, 7 | 1101 | |||
BC065563.1 | - | 6, 7 | 1088 | 6144541 |