Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018463.3 | NP_060933.3 | 1, 2 | 225 | ||
NR_130744.1 | 1, 2 | 224 | ||||
XM_005253707.4 | XP_005253764.1 | 1, 2 | 194 | |||
GenBank mRNA | AF220048.1 | - | 479 | |||
AK027483.1 | - | 1, 2 | 149 | |||
AK130319.1 | - | 1, 2 | 165 | |||
AK291110.1 | - | 1, 2 | 153 | |||
AK300905.1 | - | 1, 2 | 153 | |||
AY599883.1 | - | 1, 2 | 85 | |||
BC006552.2 | - | 1, 2 | 110 | 3029909 | ||
BC013399.2 | - | 1, 2 | 110 | 3030623 | ||
BI261823.1 | - | 1, 2 | 225 | |||
BQ058412.1 | - | 224 | ||||
KU178719.1 | - | 1, 2 | 85 | |||
KU178720.1 | - | 1, 2 | 85 | |||
KU178721.1 | - | 1, 2 | 85 |