Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018463.3 | NP_060933.3 | 9 | 1049 | ||
NR_130744.1 | 9 | 1124 | ||||
XM_005253707.4 | XP_005253764.1 | 9 | 1018 | |||
XM_005253709.4 | XP_005253766.1 | 6 | 692 | |||
XM_017019641.1 | XP_016875130.1 | 6 | 663 | |||
GenBank mRNA | AK027483.1 | - | 9 | 973 | ||
AK291110.1 | - | 9 | 977 | |||
AK297189.1 | - | 6 | 675 | |||
AK300905.1 | - | 9 | 1053 | |||
BC006552.2 | - | 9 | 934 | 3029909 | ||
BC013399.2 | - | 9 | 934 | 3030623 | ||
KU178719.1 | - | 9 | 909 | |||
KU178720.1 | - | 9 | 909 | |||
KU178721.1 | - | 9 | 849 | |||
KU178722.1 | - | 6 | 599 |