Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018464.4 | NP_060934.1 | 3 | 466 | ||
XM_017016409.1 | XP_016871898.1 | 4 | 707 | |||
GenBank mRNA | AF220049.1 | - | 3 | 405 | ||
AK312017.1 | - | 3 | 341 | |||
AY960578.1 | - | 3 | 349 | |||
BC005962.1 | - | 3 | 442 | 4051044 | ||
BC007043.1 | - | 3 | 398 | 3930701 | ||
BC008474.1 | - | 3 | 403 | 4283253 | ||
BC059168.1 | - | 3 | 413 | 4132410 | ||
DA417986.1 | - | 3 | 466 |