Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288732.1 | NP_001275661.1 | 4 | 2173 | ||
NM_001288733.1 | NP_001275662.1 | 4 | 2165 | |||
NM_018469.4 | NP_060939.3 | 4 | 2173 | |||
XM_011524998.1 | XP_011523300.1 | 4 | 2162 | |||
XM_011524999.1 | XP_011523301.1 | 4 | 2086 | |||
XM_011525000.1 | XP_011523302.1 | 4 | 2143 | |||
XM_017024846.1 | XP_016880335.1 | 4 | 2143 | |||
GenBank mRNA | AB051525.2 | - | 3 | 1992 | ||
AF163260.1 | - | 2 | 210 | |||
AK000758.1 | - | 3 | 409 | |||
AK091156.1 | - | 3 | 1162 | |||
AK091214.1 | - | 4 | 2090 | |||
AL832371.1 | - | 4 | 2073 | |||
AL834251.2 | - | 4 | 2084 | |||
BC008896.2 | - | 3 | 1350 | |||
BC036672.1 | - | 4 | 2144 | |||
BC040521.1 | - | 4 | 2077 | 4837394 | ||
BC040522.1 | - | 4 | 2146 | |||
BC108279.1 | - | 2 | 231 | |||
CR627433.1 | - | 4 | 2166 |