Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048221.2 | NP_001041686.1 | 3, 4 | 482 | ||
NM_001048223.2 | NP_001041688.1 | 3, 4 | 431 | |||
NM_001048225.2 | NP_001041690.2 | 2, 3 | 757 | |||
NM_001197139.1 | NP_001184068.1 | 3, 4 | 315 | |||
NM_001197140.1 | NP_001184069.1 | 3, 4 | 361 | |||
NM_018478.3 | NP_060948.3 | 3, 4 | 803 | |||
GenBank mRNA | AJ276469.1 | - | 3, 4 | 400 | ||
AL591565.1 | - | 3, 4 | 315 | |||
BP330715.1 | - | 3, 4 | 482 |
Gene Symbol: SYS1-DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003189.2 | 5, 6 | 666 | |||
GenBank mRNA | BC023562.2 | - | 5, 6 | 635 |