Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258338.1 | NP_001245267.1 | 3 | 315 | ||
NM_018838.4 | NP_061326.1 | 4 | 403 | |||
GenBank mRNA | AF112208.1 | - | 4 | 423 | ||
AF217092.1 | - | 4 | 382 | |||
AK222619.1 | - | 4 | 390 | |||
BC005936.1 | - | 4 | 392 | 4069306 | ||
BG943528.1 | - | 4 | 388 | |||
BM548072.1 | - | 470 | ||||
BT007220.1 | - | 4 | 370 | |||
BU532909.1 | - | 3 | 302 | |||
DW441120.1 | - | 3 | 329 | |||
U34343.1 | - | 509 |