Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199534.1 | NP_001186463.1 | 2 | 376 | ||
NM_018840.4 | NP_061328.1 | 2 | 376 | |||
NM_199483.2 | NP_955777.1 | 2 | 376 | |||
NR_026562.3 | 2 | 376 | ||||
GenBank mRNA | AF112213.1 | - | 2 | 340 | ||
AF274936.1 | - | 2 | 271 | |||
AF274948.1 | - | 2 | 189 | |||
BC001871.1 | - | 2 | 293 | 3546888 | ||
BC004446.1 | - | 2 | 301 | 2819807 | ||
BG395753.1 | - | 2 | 347 | |||
BI223439.1 | - | 2 | 380 | |||
BQ086248.1 | - | 2 | 325 | |||
BU174619.1 | - | 344 | ||||
BU189639.1 | - | 2 | 288 | |||
BU194101.1 | - | 303 | ||||
BU848918.1 | - | 2 | 335 | |||
S83364.1 | - | 2 | 116 |
Gene Symbol: TGIF2-C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199535.1 | NP_001186464.1 | 3 | 352 |