Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002749.3 | NP_002740.2 | 4 | 739 | ||
NM_139032.2 | NP_620601.1 | 2 | 547 | |||
NM_139033.2 | NP_620602.2 | 4 | 950 | |||
NM_139034.2 | NP_620603.2 | 4 | 651 | |||
XM_006721557.3 | XP_006721620.1 | 5 | 715 | |||
XM_006721558.3 | XP_006721621.1 | 5 | 968 | |||
XM_006721559.3 | XP_006721622.1 | 5 | 877 | |||
XM_011523957.2 | XP_011522259.1 | 2 | 454 | |||
GenBank mRNA | AB209611.1 | - | 1 | 2245 | ||
AK295376.1 | - | 4 | 644 | |||
AY534741.1 | - | 3 | 577 | |||
BC007404.2 | - | 2 | 515 | 3009873 | ||
BC007992.2 | - | 2 | 515 | 3009873 | ||
BC009963.2 | - | 4 | 671 | 4300124 | ||
BC030134.1 | - | 4 | 586 | 4111084 | ||
U25278.1 | - | 4 | 644 | |||
U29725.1 | - | 4 | 785 | |||
U29726.1 | - | 4 | 618 |