Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021570.3 | NP_067545.3 | 1 | 257 | ||
GenBank mRNA | BC064363.1 | - | 1 | 302 | 5555949 |