Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018920.3 | NP_061743.1 | 1 | 2221 | ||
NM_032087.2 | NP_114476.1 | 1 | 2221 | |||
GenBank mRNA | AB208958.1 | - | 1 | 1951 | ||
AF152327.1 | - | 1 | 2060 | |||
AF152514.1 | - | 1 | 2060 | |||
BC136747.1 | - | 1 | 2125 | 9020737 | ||
BC136748.1 | - | 1 | 2125 | 9020738 |