Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135651.2 | NP_001129123.1 | 3 | 353 | ||
NM_001135652.2 | NP_001129124.1 | 1 | 46 | |||
NM_002759.3 | NP_002750.1 | 3 | 587 | |||
XM_011532987.2 | XP_011531289.1 | 3 | 287 | |||
XM_017004503.1 | XP_016859992.1 | 3 | 283 | |||
GenBank mRNA | AK290655.1 | - | 3 | 273 | ||
AK313818.1 | - | 2 | 110 | |||
AY302136.1 | - | 1 | 30 | |||
BC007769.2 | - | 2 | 129 | |||
BC022314.1 | - | 3 | 241 | |||
BC040851.1 | - | 3 | 353 | |||
BC057805.1 | - | 3 | 258 | |||
BC093676.1 | - | 1 | 58 | 7939521 | ||
BC101475.1 | - | 1 | 58 | 8068981 | ||
BC143228.1 | - | 1 | 57 | |||
BC143230.1 | - | 1 | 58 | 9051731 | ||
M35663.1 | - | 2 | 216 | |||
M85294.1 | - | 1 | 60 |