Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004705.3 | NP_004696.2 | 2 | 299 | ||
NR_130898.1 | 2 | 299 | ||||
GenBank mRNA | AF007393.1 | - | 2 | 170 | ||
AF081567.1 | - | 2 | 170 | |||
AK291843.1 | - | 2 | 171 | |||
BC110304.1 | - | 2 | 276 | 5222582 | ||
BC117138.1 | - | 2 | 228 | 40125689 | ||
BC117140.1 | - | 2 | 228 | 40125691 | ||
DR005363.1 | - | 2 | 305 |