Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPCPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019593.3 | NP_062539.1 | 2 | 226 | ||
XM_005260758.4 | XP_005260815.1 | 3 | 377 | |||
XM_005260761.1 | XP_005260818.1 | 2 | 226 | |||
XM_006723596.3 | XP_006723659.1 | 3 | 1150 | |||
GenBank mRNA | AB037855.1 | - | 2 | 217 | ||
AK308111.1 | - | 2 | 224 | |||
BC027588.2 | - | 2 | 225 | 4827346 | ||
BI561344.1 | - | 2 | 160 |