Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR45B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019613.3 | NP_062559.2 | 7 | 880 | ||
XM_005256377.4 | XP_005256434.1 | 6 | 703 | |||
XM_017024862.1 | XP_016880351.1 | 8 | 2217 | |||
XM_017024863.1 | XP_016880352.1 | 9 | 1015 | |||
GenBank mRNA | AF091083.1 | - | 5 | 523 | ||
AK297477.1 | - | 7 | 634 | |||
AM182326.1 | - | 7 | 685 | |||
AY691427.1 | - | 5 | 511 | |||
BC000974.2 | - | 7 | 629 | |||
BC007838.1 | - | 7 | 852 | |||
CR456770.1 | - | 5 | 511 |