Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019617.3 | NP_062563.3 | 3 | 244 | ||
GenBank mRNA | AB039886.1 | - | 3 | 244 | ||
AJ966789.1 | - | 3 | 244 | |||
AK098489.1 | - | 1 | 3074 | |||
AY139184.1 | - | 3 | 182 | |||
AY359050.1 | - | 3 | 168 | |||
AY456961.1 | - | 3 | 181 | |||
BC059778.1 | - | 3 | 170 |