Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160130.1 | NP_001153602.1 | 11 | 1977 | ||
NM_001160132.1 | NP_001153604.1 | 12 | 2034 | |||
NM_001160133.1 | NP_001153605.1 | 13 | 2061 | |||
NM_001160134.1 | NP_001153606.1 | 10 | 1674 | |||
NM_019842.3 | NP_062816.2 | 12 | 2004 | |||
GenBank mRNA | AF202977.1 | - | 12 | 1552 | ||
AF249278.1 | - | 12 | 1766 | |||
AF263835.1 | - | 12 | 1444 | |||
AK299550.1 | - | 10 | 1274 | |||
AK299741.1 | - | 3 | 216 | |||
AK316070.1 | - | 3 | 218 | |||
BC117359.1 | - | 11 | 1648 | 40125910 | ||
BC143554.1 | - | 11 | 1648 | 9052065 | ||
BC143555.1 | - | 10 | 1521 |