Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031849.2 | NP_001027019.1 | 4 | 872 | ||
NM_001879.5 | NP_001870.3 | 4 | 872 | |||
NM_139125.3 | NP_624302.1 | 4 | 872 | |||
NR_033519.1 | 3 | 640 | ||||
XM_006713701.2 | XP_006713764.1 | 5 | 945 | |||
XM_011512989.2 | XP_011511291.1 | 4 | 866 | |||
XM_011512990.2 | XP_011511292.1 | 4 | 866 | |||
XM_011512991.2 | XP_011511293.1 | 4 | 861 | |||
XM_017006869.1 | XP_016862358.1 | 5 | 950 | |||
XM_017006870.1 | XP_016862359.1 | 3 | 838 | |||
XM_017006871.1 | XP_016862360.1 | 4 | 866 | |||
XM_017006872.1 | XP_016862361.1 | 4 | 725 | |||
GenBank mRNA | AF284421.1 | - | 4 | 572 | ||
AK129893.1 | - | 4 | 571 | |||
AK291157.1 | - | 4 | 708 | |||
AK291686.1 | - | 4 | 550 | |||
AK304334.1 | - | 3 | 350 | |||
BC039724.1 | - | 4 | 807 | |||
BC106945.1 | - | 4 | 532 | 40034551 | ||
BC106946.1 | - | 4 | 532 | 40034552 | ||
BX647237.1 | - | 3 | 355 | |||
CB162101.1 | - | 2 | 97 | |||
CR749615.1 | - | 4 | 774 |