Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020137.4 | NP_064522.3 | 12 | 968 | ||
XM_011543935.1 | XP_011542237.1 | 12 | 968 | |||
XM_011543936.1 | XP_011542238.1 | 12 | 968 | |||
XM_011543937.1 | XP_011542239.1 | 12 | 1015 | |||
XM_017029656.1 | XP_016885145.1 | 12 | 968 | |||
XM_017029657.1 | XP_016885146.1 | 12 | 1339 | |||
XM_017029658.1 | XP_016885147.1 | 12 | 2278 | |||
XM_017029659.1 | XP_016885148.1 | 12 | 1015 | |||
XM_017029660.1 | XP_016885149.1 | 12 | 1369 | |||
XM_017029661.1 | XP_016885150.1 | 12 | 1015 | |||
GenBank mRNA | AB032993.1 | - | 12 | 913 | ||
AK292158.1 | - | 12 | 939 | |||
BC001522.2 | - | 2 | 81 | |||
BC101544.1 | - | 12 | 966 | 8069050 | ||
BC101546.1 | - | 12 | 966 | 8069052 | ||
BX640704.1 | - | 6 | 652 | |||
BX647801.1 | - | 11 | 808 | |||
BX647804.1 | - | 11 | 808 |