Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253723.1 | NP_001240652.1 | 8, 9 | 1020 | ||
NM_001253724.1 | NP_001240653.1 | 8, 9 | 1023 | |||
NM_020134.3 | NP_064519.2 | 8, 9 | 1103 | |||
GenBank mRNA | AF157634.1 | - | 7, 8 | 945 | ||
AF264015.1 | - | 8, 9 | 1107 | |||
AJ251275.1 | - | 7, 8 | 945 | |||
AL713706.1 | - | 6, 7 | 645 | |||
BC002874.2 | - | 8, 9 | 1020 | 3940393 | ||
BT006871.1 | - | 7, 8 | 945 |