Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020139.3 | NP_064524.3 | 9, 10 | 787 | ||
XM_005263140.3 | XP_005263197.1 | 8, 9 | 668 | |||
XM_006714274.3 | XP_006714337.1 | 10, 11 | 824 | |||
XM_011532127.2 | XP_011530429.1 | 10, 11 | 866 | |||
XM_011532128.2 | XP_011530430.1 | 9, 10 | 824 | |||
XM_017008461.1 | XP_016863950.1 | 10, 11 | 863 | |||
XM_017008462.1 | XP_016863951.1 | 9, 10 | 821 | |||
GenBank mRNA | AF164790.1 | - | 9, 10 | 751 | ||
AK023323.1 | - | 8, 9 | 1760 | |||
AK128463.1 | - | 9, 10 | 2502 | |||
AK290160.1 | - | 9, 10 | 743 | |||
AK300626.1 | - | 9, 10 | 632 | |||
AY358841.1 | - | 9, 10 | 753 | |||
BC001953.1 | - | 9, 10 | 780 | 2905279 | ||
BC037277.2 | - | 8, 9 | 1766 | 4838679 |