Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128626.1 | NP_001122098.1 | 4 | 734 | ||
NM_001128627.1 | NP_001122099.1 | 4 | 432 | |||
NM_020148.2 | NP_064533.3 | 4 | 734 | |||
XM_005258122.4 | XP_005258179.1 | 4 | 917 | |||
XM_011525701.1 | XP_011524003.1 | 4 | 923 | |||
XM_011525702.1 | XP_011524004.1 | 4 | 435 | |||
XM_011525703.2 | XP_011524005.1 | 4 | 917 | |||
GenBank mRNA | AB032961.2 | - | 4 | 786 | ||
AJ277587.1 | - | 2 | 321 | |||
AK290180.1 | - | 2 | 292 | |||
AK301711.1 | - | 3 | 201 | |||
AL833817.1 | - | 5 | 412 | |||
BC115005.1 | - | 4 | 559 | 7782242 | ||
BC125206.1 | - | 2 | 248 | 40123468 | ||
BC125207.1 | - | 2 | 248 | 40123469 | ||
DB056921.1 | - | 4 | 432 |