Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002795.3 | NP_002786.2 | 3, 4 | 399 | ||
GenBank mRNA | BC013008.2 | - | 3, 4 | 362 | 3010175 | |
D26598.1 | - | 3, 4 | 302 |