Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002796.2 | NP_002787.2 | 2 | 331 | ||
GenBank mRNA | AK294149.1 | - | 2 | 333 | ||
AK313825.1 | - | 2 | 333 | |||
BC000331.1 | - | 2 | 322 | |||
BC008314.2 | - | 2 | 330 | |||
BC010088.2 | - | 2 | 330 | |||
BC010098.2 | - | 2 | 330 | |||
BC011768.2 | - | 2 | 330 | |||
BC012168.2 | - | 2 | 330 | |||
BC017307.1 | - | 2 | 380 | |||
BC017451.2 | - | 2 | 330 | |||
BC017486.2 | - | 2 | 330 | |||
BT006917.1 | - | 2 | 308 | |||
CR456820.1 | - | 2 | 308 | |||
D26600.1 | - | 2 | 331 | |||
S71381.1 | - | 2 | 314 |